siRNA Details
| Gene Symbol: Hoxb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100787.1 | NP_001094257.1 | 2 | 1731 | ||
| GenBank mRNA | BC161863.1 | - | 2 | 1731 | 7098209 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxb4| Gene Symbol: Hoxb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100787.1 | NP_001094257.1 | 2 | 1731 | ||
| GenBank mRNA | BC161863.1 | - | 2 | 1731 | 7098209 | |