siRNA Details
Gene Symbol: Coa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109047.1 | NP_001102517.1 | 2 | 654 | ||
GenBank mRNA | FQ221438.1 | - | 2 | 660 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Coa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109047.1 | NP_001102517.1 | 2 | 654 | ||
GenBank mRNA | FQ221438.1 | - | 2 | 660 |