siRNA Details
| Gene Symbol: NOLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284389.1 | NP_001271318.1 | 6, 7 | 963 | ||
| NM_004741.4 | NP_004732.2 | 6, 7 | 960 | |||
| GenBank mRNA | AK056755.1 | - | 5, 6 | 583 | ||
| AK225101.1 | - | 6, 7 | 750 | |||
| AK314363.1 | - | 6, 7 | 747 | |||
| AY820769.1 | - | 6, 7 | 728 | |||
| BC001883.1 | - | 6, 7 | 735 | 3533568 | ||
| BC006769.2 | - | 2, 3 | 245 | 2900024 | ||
| L37710.1 | - | 1, 2 | 44 | |||
| Z34289.1 | - | 6, 7 | 779 | |||

NOLC1
Genomic Map