siRNA Details
| Gene Symbol: RGD1565533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134601.2 | NP_001128073.2 | 4 | 441 | ||
| XM_006247404.1 | XP_006247466.1 | 4 | 465 | |||
| XM_006247405.2 | XP_006247467.1 | 4 | 465 | |||
| XM_008768106.1 | XP_008766328.1 | 4 | 465 | |||

RGD1565533
Genomic Map