siRNA Details
| Gene Symbol: RGD1565033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109050.1 | NP_001102520.1 | 2, 3 | 354 | ||
| XM_006247636.1 | XP_006247698.1 | 3, 4 | 1161 | |||
| XM_006247637.1 | XP_006247699.1 | 3, 4 | 1158 | |||
| XM_006247638.1 | XP_006247700.1 | 4, 5 | 1038 | |||
| XM_008768383.1 | XP_008766605.1 | 4, 5 | 816 | |||

RGD1565033
Genomic Map