siRNA Details
| Gene Symbol: RGD1565033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109050.1 | NP_001102520.1 | 2 | 259 | ||
| XM_006247636.1 | XP_006247698.1 | 3 | 1066 | |||
| XM_006247637.1 | XP_006247699.1 | 3 | 1063 | |||
| XM_006247638.1 | XP_006247700.1 | 4 | 943 | |||
| XM_006247639.2 | XP_006247701.1 | 3 | 1098 | |||
| XM_006247640.2 | XP_006247702.1 | 3 | 1070 | |||
| XM_008768383.1 | XP_008766605.1 | 4 | 721 | |||
| GenBank mRNA | BC158779.1 | - | 2 | 248 | 7134339 | |

RGD1565033
Genomic Map