siRNA Details
| Gene Symbol: Tex19.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017482.1 | NP_001017482.1 | 3 | 748 | ||
| GenBank mRNA | BC083916.1 | - | 3 | 748 | 7134696 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex19.1| Gene Symbol: Tex19.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017482.1 | NP_001017482.1 | 3 | 748 | ||
| GenBank mRNA | BC083916.1 | - | 3 | 748 | 7134696 | |