siRNA Details
| Gene Symbol: Lca5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017484.1 | NP_001017484.1 | 6 | 1126 | ||
| XM_006248178.2 | XP_006248240.1 | 7 | 1352 | |||
| XM_006248179.2 | XP_006248241.1 | 8 | 1454 | |||
| XM_006248180.2 | XP_006248242.1 | 7 | 1401 | |||
| XM_006248181.2 | XP_006248243.1 | 7 | 1292 | |||
| XM_006248182.2 | XP_006248244.1 | 6 | 1190 | |||
| GenBank mRNA | BC082082.1 | - | 6 | 1126 | 7133412 | |

Lca5l
Genomic Map