siRNA Details
| Gene Symbol: Lca5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017484.1 | NP_001017484.1 | 7, 8 | 1275 | ||
| XM_006248178.2 | XP_006248240.1 | 8, 9 | 1501 | |||
| XM_006248179.2 | XP_006248241.1 | 9, 10 | 1603 | |||
| XM_006248180.2 | XP_006248242.1 | 8, 9 | 1550 | |||
| XM_006248181.2 | XP_006248243.1 | 8, 9 | 1441 | |||
| XM_006248182.2 | XP_006248244.1 | 7, 8 | 1339 | |||
| GenBank mRNA | BC082082.1 | - | 7, 8 | 1275 | 7133412 | |

Lca5l
Genomic Map