siRNA Details
| Gene Symbol: Cd96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025032.1 | NP_001020203.1 | 13 | 1537 | ||
| XM_008768647.1 | XP_008766869.1 | 13 | 1741 | |||
| GenBank mRNA | BC091206.1 | - | 13 | 1537 | 7380788 | |
| FQ228851.1 | - | 13 | 1525 | |||

Cd96
Genomic Map