siRNA Details
| Gene Symbol: Itgb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147139.2 | NP_671480.2 | 3 | 243 | ||
| GenBank mRNA | CB731093.1 | - | 3 | 213 | ||
| DV216421.1 | - | 3 | 425 | |||
| EV772196.1 | - | 2 | 96 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itgb5| Gene Symbol: Itgb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147139.2 | NP_671480.2 | 3 | 243 | ||
| GenBank mRNA | CB731093.1 | - | 3 | 213 | ||
| DV216421.1 | - | 3 | 425 | |||
| EV772196.1 | - | 2 | 96 | |||