siRNA Details
| Gene Symbol: Itgb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147139.2 | NP_671480.2 | 10 | 1482 | ||
| GenBank mRNA | DY316865.1 | - | 2 | 226 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itgb5| Gene Symbol: Itgb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147139.2 | NP_671480.2 | 10 | 1482 | ||
| GenBank mRNA | DY316865.1 | - | 2 | 226 | ||