siRNA Details
| Gene Symbol: LOC100911042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751070.3 | XP_003751118.1 | 3 | 587 | ||
| Gene Symbol: Smco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248472.2 | XP_006248534.1 | 589 | |||
| GenBank mRNA | FQ217149.1 | - | 587 | |||

LOC100911042
Genomic Map