siRNA Details
| Gene Symbol: Wdr53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109055.1 | NP_001102525.1 | 3 | 883 | ||
| XM_006248459.2 | XP_006248521.1 | 3 | 986 | |||
| XM_006248460.2 | XP_006248522.1 | 3 | 787 | |||
| XM_006248461.2 | XP_006248523.1 | 3 | 621 | |||

Wdr53
Genomic Map