siRNA Details
| Gene Symbol: Yeats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109057.1 | NP_001102527.1 | 25 | 3519 | ||
| XM_008768826.1 | XP_008767048.1 | 19 | 3189 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yeats2| Gene Symbol: Yeats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109057.1 | NP_001102527.1 | 25 | 3519 | ||
| XM_008768826.1 | XP_008767048.1 | 19 | 3189 | |||