siRNA Details
| Gene Symbol: Yeats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109057.1 | NP_001102527.1 | 26 | 3566 | ||
| XM_008768826.1 | XP_008767048.1 | 20 | 3236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yeats2| Gene Symbol: Yeats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109057.1 | NP_001102527.1 | 26 | 3566 | ||
| XM_008768826.1 | XP_008767048.1 | 20 | 3236 | |||