siRNA Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768898.1 | XP_008767120.1 | 4 | 403 | ||
| XM_008768899.1 | XP_008767121.1 | 4 | 403 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spidr| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768898.1 | XP_008767120.1 | 4 | 403 | ||
| XM_008768899.1 | XP_008767121.1 | 4 | 403 | |||