siRNA Details
| Gene Symbol: Mcemp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134602.1 | NP_001128074.1 | 4, 5 | 348 | ||
| XM_008768987.1 | XP_008767209.1 | 7, 8 | 669 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcemp1| Gene Symbol: Mcemp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134602.1 | NP_001128074.1 | 4, 5 | 348 | ||
| XM_008768987.1 | XP_008767209.1 | 7, 8 | 669 | |||