siRNA Details
| Gene Symbol: Stard13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109060.1 | NP_001102530.1 | 3, 4 | 426 | ||
| XM_006248743.2 | XP_006248805.1 | 3, 4 | 646 | |||
| XM_006248749.2 | XP_006248811.1 | 3, 4 | 1264 | |||
| GenBank mRNA | KF112845.1 | - | 3, 4 | 332 | ||
| KF112846.1 | - | 3, 4 | 332 | |||

Stard13
Genomic Map