siRNA Details
| Gene Symbol: Tex26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134603.1 | NP_001128075.1 | 5 | 576 | ||
| XM_006248811.2 | XP_006248873.1 | 5 | 715 | |||
| XM_006248813.2 | XP_006248875.1 | 5 | 881 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex26| Gene Symbol: Tex26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134603.1 | NP_001128075.1 | 5 | 576 | ||
| XM_006248811.2 | XP_006248873.1 | 5 | 715 | |||
| XM_006248813.2 | XP_006248875.1 | 5 | 881 | |||