siRNA Details
Gene Symbol: Cdk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109061.1 | NP_001102531.1 | 4 | 349 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
