siRNA Details
| Gene Symbol: Zkscan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025760.1 | NP_001020931.1 | 6 | 1711 | ||
| XM_006249015.2 | XP_006249077.1 | 4 | 1535 | |||
| GenBank mRNA | BC099191.1 | - | 6 | 1711 | 7383463 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zkscan1| Gene Symbol: Zkscan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025760.1 | NP_001020931.1 | 6 | 1711 | ||
| XM_006249015.2 | XP_006249077.1 | 4 | 1535 | |||
| GenBank mRNA | BC099191.1 | - | 6 | 1711 | 7383463 | |