siRNA Details
| Gene Symbol: Gal3st4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109064.1 | NP_001102534.1 | 3 | 576 | ||
| XM_006249025.2 | XP_006249087.1 | 3 | 808 | |||
| XM_006249026.2 | XP_006249088.1 | 3 | 594 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gal3st4| Gene Symbol: Gal3st4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109064.1 | NP_001102534.1 | 3 | 576 | ||
| XM_006249025.2 | XP_006249087.1 | 3 | 808 | |||
| XM_006249026.2 | XP_006249088.1 | 3 | 594 | |||