siRNA Details
| Gene Symbol: Mapkapk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025761.2 | NP_001020932.2 | 10, 11 | 1129 | ||
| NM_001164043.1 | NP_001157515.1 | 13, 14 | 1498 | |||
| GenBank mRNA | BC099161.1 | - | 10, 11 | 1129 | 7381631 | |

Mapkapk5
Genomic Map