siRNA Details
| Gene Symbol: Cyp3a85-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769056.1 | XP_008767278.1 | 7 | 692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp3a85-ps| Gene Symbol: Cyp3a85-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769056.1 | XP_008767278.1 | 7 | 692 | ||