siRNA Details
Gene Symbol: Tnfrsf11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271235.1 | NP_001258164.1 | 3 | 260 | ||
XM_008769487.1 | XP_008767709.1 | 3 | 243 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tnfrsf11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271235.1 | NP_001258164.1 | 3 | 260 | ||
XM_008769487.1 | XP_008767709.1 | 3 | 243 |