siRNA Details
| Gene Symbol: LOC498222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100788.1 | NP_001094258.1 | 4 | 947 | ||
| XM_006249714.2 | XP_006249776.1 | 5 | 3209 | |||
| XM_008769488.1 | XP_008767710.1 | 4 | 1072 | |||
| XM_008769489.1 | XP_008767711.1 | 3 | 2917 | |||
| GenBank mRNA | BC099745.1 | - | 1 | 203 | ||

LOC498222
Genomic Map