siRNA Details
| Gene Symbol: LOC498231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047925.1 | NP_001041390.1 | 4 | 379 | ||
| GenBank mRNA | AY539895.1 | - | 4 | 379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC498231| Gene Symbol: LOC498231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047925.1 | NP_001041390.1 | 4 | 379 | ||
| GenBank mRNA | AY539895.1 | - | 4 | 379 | ||