siRNA Details
| Gene Symbol: Lax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017491.1 | NP_001017491.1 | 6 | 1128 | ||
| XM_006249793.2 | XP_006249855.1 | 6 | 1263 | |||
| GenBank mRNA | BC089837.1 | - | 6 | 1128 | 7365271 | |
| FQ234558.1 | - | 1 | 75 | |||

Lax1
Genomic Map