siRNA Details
| Gene Symbol: LOC498236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047926.1 | NP_001041391.1 | 9 | 820 | ||
| XM_008769565.1 | XP_008767787.1 | 10 | 1019 | |||
| GenBank mRNA | AY539937.1 | - | 9 | 820 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC498236| Gene Symbol: LOC498236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047926.1 | NP_001041391.1 | 9 | 820 | ||
| XM_008769565.1 | XP_008767787.1 | 10 | 1019 | |||
| GenBank mRNA | AY539937.1 | - | 9 | 820 | ||