siRNA Details
| Gene Symbol: PNMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006029.4 | NP_006020.4 | 1 | 1423 | ||
| GenBank mRNA | AF320308.1 | - | 1 | 908 | ||
| AK290980.1 | - | 1 | 760 | |||
| AL834327.1 | - | 1 | 156 | |||
| BC039577.1 | - | 1 | 726 | 4431875 | ||
| CR457112.1 | - | 1 | 637 | |||

PNMA1
Genomic Map