siRNA Details
| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017494.1 | NP_001017494.1 | 4 | 780 | ||
| GenBank mRNA | BC079114.1 | - | 4 | 780 | 7126053 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Blzf1| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017494.1 | NP_001017494.1 | 4 | 780 | ||
| GenBank mRNA | BC079114.1 | - | 4 | 780 | 7126053 | |