siRNA Details
| Gene Symbol: Dusp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191929.1 | NP_001178858.1 | 5 | 3060 | ||
| XM_006250187.2 | XP_006250249.1 | 7 | 3260 | |||
| XM_006250188.2 | XP_006250250.1 | 7 | 3256 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp27| Gene Symbol: Dusp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191929.1 | NP_001178858.1 | 5 | 3060 | ||
| XM_006250187.2 | XP_006250249.1 | 7 | 3260 | |||
| XM_006250188.2 | XP_006250250.1 | 7 | 3256 | |||