siRNA Details
| Gene Symbol: Fcrlb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191931.1 | NP_001178860.1 | 6 | 1215 | ||
| XM_008769747.1 | XP_008767969.1 | 4 | 1399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fcrlb| Gene Symbol: Fcrlb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191931.1 | NP_001178860.1 | 6 | 1215 | ||
| XM_008769747.1 | XP_008767969.1 | 4 | 1399 | |||