siRNA Details
| Gene Symbol: Slamf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191932.1 | NP_001178861.1 | 2, 3 | 444 | ||
| XM_006250301.2 | XP_006250363.1 | 2, 3 | 865 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slamf6| Gene Symbol: Slamf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191932.1 | NP_001178861.1 | 2, 3 | 444 | ||
| XM_006250301.2 | XP_006250363.1 | 2, 3 | 865 | |||