siRNA Details
| Gene Symbol: Kif26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109079.2 | NP_001102549.2 | 3 | 989 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif26b| Gene Symbol: Kif26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109079.2 | NP_001102549.2 | 3 | 989 | ||