siRNA Details
| Gene Symbol: Cnst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250411.2 | XP_006250473.1 | 2 | 1170 | ||
| XM_006250412.2 | XP_006250474.1 | 2 | 559 | |||
| XM_006250413.2 | XP_006250475.1 | 2 | 559 | |||
| XM_006250414.2 | XP_006250476.1 | 284 | ||||
| XM_573525.5 | XP_573525.5 | 2 | 559 | |||
| Gene Symbol: LOC100912475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250360.2 | XP_006250422.1 | 2 | 559 | ||

Cnst
Genomic Map