siRNA Details
Gene Symbol: Bmp2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770127.1 | XP_008768349.1 | 14 | 2181 | ||
GenBank mRNA | BC098765.1 | - | 2 | 140 | ||
BC103651.1 | - | 6 | 976 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bmp2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770127.1 | XP_008768349.1 | 14 | 2181 | ||
GenBank mRNA | BC098765.1 | - | 2 | 140 | ||
BC103651.1 | - | 6 | 976 |