siRNA Details
| Gene Symbol: Srp72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170601.1 | NP_001164072.1 | 528 | |||
| XM_008770133.1 | XP_008768355.1 | 1166 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srp72| Gene Symbol: Srp72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170601.1 | NP_001164072.1 | 528 | |||
| XM_008770133.1 | XP_008768355.1 | 1166 | ||||