siRNA Details
| Gene Symbol: Srp72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170601.1 | NP_001164072.1 | 1223 | |||
| GenBank mRNA | BC107914.1 | - | 373 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srp72| Gene Symbol: Srp72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170601.1 | NP_001164072.1 | 1223 | |||
| GenBank mRNA | BC107914.1 | - | 373 | |||