siRNA Details
| Gene Symbol: Pdcl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_573585.6 | XP_573585.3 | 6 | 956 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdcl2| Gene Symbol: Pdcl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_573585.6 | XP_573585.3 | 6 | 956 | ||