siRNA Details
| Gene Symbol: Shisa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109087.2 | NP_001102557.1 | 2 | 955 | ||
| GenBank mRNA | BC168960.1 | - | 2 | 955 | 9023733 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shisa3| Gene Symbol: Shisa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109087.2 | NP_001102557.1 | 2 | 955 | ||
| GenBank mRNA | BC168960.1 | - | 2 | 955 | 9023733 | |