siRNA Details
| Gene Symbol: Slc30a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109088.1 | NP_001102558.1 | 11 | 981 | ||
| XM_006250963.1 | XP_006251025.1 | 11 | 1051 | |||
| XM_006250965.2 | XP_006251027.1 | 11 | 1451 | |||
| GenBank mRNA | FQ222791.1 | - | 9 | 1274 | ||

Slc30a9
Genomic Map