siRNA Details
| Gene Symbol: Fam114a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250991.2 | XP_006251053.1 | 14 | 2189 | ||
| XM_008770176.1 | XP_008768398.1 | 14 | 2477 | |||
| XM_008770177.1 | XP_008768399.1 | 14 | 2195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam114a1| Gene Symbol: Fam114a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250991.2 | XP_006251053.1 | 14 | 2189 | ||
| XM_008770176.1 | XP_008768398.1 | 14 | 2477 | |||
| XM_008770177.1 | XP_008768399.1 | 14 | 2195 | |||