siRNA Details
| Gene Symbol: LOC498368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017500.1 | NP_001017500.1 | 2 | 867 | ||
| GenBank mRNA | BC079124.1 | - | 2 | 867 | 7126862 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC498368| Gene Symbol: LOC498368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017500.1 | NP_001017500.1 | 2 | 867 | ||
| GenBank mRNA | BC079124.1 | - | 2 | 867 | 7126862 | |