siRNA Details
Gene Symbol: GPR50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004224.3 | NP_004215.2 | 2 | 508 | ||
GenBank mRNA | AY114101.1 | - | 2 | 220 | ||
BC103696.1 | - | 2 | 418 | 40006713 | ||
BC105683.1 | - | 2 | 418 | 40006718 | ||
BC105684.1 | - | 2 | 418 | 40006715 | ||
BP245442.1 | - | 2 | 508 | |||
EU432118.1 | - | 2 | 370 | |||
U52219.1 | - | 2 | 439 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
