siRNA Details
| Gene Symbol: Msantd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109089.1 | NP_001102559.1 | 2 | 583 | ||
| XM_006251345.2 | XP_006251407.1 | 3 | 2107 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msantd1| Gene Symbol: Msantd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109089.1 | NP_001102559.1 | 2 | 583 | ||
| XM_006251345.2 | XP_006251407.1 | 3 | 2107 | |||