siRNA Details
| Gene Symbol: Ccdc117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017502.1 | NP_001017502.1 | 3, 4 | 488 | ||
| GenBank mRNA | BC087119.1 | - | 3, 4 | 488 | 7097247 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc117| Gene Symbol: Ccdc117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017502.1 | NP_001017502.1 | 3, 4 | 488 | ||
| GenBank mRNA | BC087119.1 | - | 3, 4 | 488 | 7097247 | |