siRNA Details
| Gene Symbol: Mrps24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077659.1 | NP_001071127.1 | 4 | 408 | ||
| GenBank mRNA | BC157807.1 | - | 4 | 404 | 8375708 | |
| BP503351.1 | - | 4 | 413 | |||
| DY319747.1 | - | 4 | 404 | |||
| FQ227686.1 | - | 4 | 426 | |||

Mrps24
Genomic Map