siRNA Details
Gene Symbol: Hus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109092.1 | NP_001102562.1 | 4 | 469 | ||
XM_006251468.1 | XP_006251530.1 | 4 | 464 | |||
XM_006251469.2 | XP_006251531.1 | 4 | 668 | |||
XM_006251470.2 | XP_006251532.1 | 4 | 460 | |||
XM_008770346.1 | XP_008768568.1 | 4 | 461 |
